missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Pyrrolidinone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 132115000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Pyrrolidinone | |
616-45-5 | |
100.0 | |
Colorless to Yellow | |
250.0°C | |
Authentic | |
Glass bottle | |
1.4860 to 1.4880 | |
500 mL | |
1.103 | |
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam | |
HNJBEVLQSNELDL-UHFFFAOYSA-N | |
pyrrolidin-2-one | |
12025 | |
CHEBI:36592 | |
99% |
98% | |
98.5 | |
25.5°C | |
1.1030g/mL | |
138°C | |
98.5% min. (GC) | |
C4H7NO | |
MFCD00005270 | |
21, 236 | |
15, 8130 | |
Solubility in water: miscible. Other solubilities: miscible with ethanol, ether, chloroform, benzene,, ethyl acetate, carbon disulfide | |
C1CC(=O)NC1 | |
85.11 | |
10.2 mPa.s (30°C) | |
85.11 | |
Liquid or Low Melting Mass |
Chemical Identifiers
616-45-5 | |
85.11 | |
HNJBEVLQSNELDL-UHFFFAOYSA-N | |
12025 | |
pyrrolidin-2-one |
C4H7NO | |
MFCD00005270 | |
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam | |
CHEBI:36592 | |
C1CC(=O)NC1 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS Signal Word: Warning
EINECSNumber : 210-483-1
RTECSNumber : UY5715000
TSCA : TSCA
Recommended Storage : Product may darken during storage
RUO – Research Use Only