Learn More
2-Pyrrolidin-2-ylpyridine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 446740010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Pyrrolidin-2-ylpyridine | |
82.0°C (0.8 mmHg) | |
Glass bottle | |
1g | |
NDCZQFDBSPOUDF-UHFFFAOYSA-N | |
2-pyrrolidin-2-ylpyridine | |
2771659 | |
95% |
77790-61-5 | |
94% min. (GC) | |
C9H12N2 | |
2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine | |
C1CC(NC1)C2=CC=CC=N2 | |
148.21 | |
148.21 |
Chemical Identifiers
77790-61-5 | |
148.21 | |
2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine | |
2-pyrrolidin-2-ylpyridine |
C9H12N2 | |
NDCZQFDBSPOUDF-UHFFFAOYSA-N | |
2771659 | |
C1CC(NC1)C2=CC=CC=N2 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
RUO â Research Use Only