missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Pyrrolidin-1-ylpyrimidin-5-yl)methanol, 97%, Maybridge

$305.40 - $305.40

Chemical Identifiers

CAS 937796-11-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H13N3O
Formula Weight 179.22
MDL Number MFCD09863240
Chemical Name or Material (2-Pyrrolidin-1-ylpyrimidin-5-yl)methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC69309CB SDS Fisher Scientific Company
CC69309CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC69309DA SDS Fisher Scientific Company
CC69309DA
Amber Glass Bottle 1g Each for $305.40
Specifications Description & Specifications

Chemical Identifiers

937796-11-7
100.0
C9H13N3O
MFCD09863240
97%
96.5
97%
179.22
(2-Pyrrolidin-1-ylpyrimidin-5-yl)methanol

Specifications

179.22
100.0
C9H13N3O
96.5
97%
97%
250mg
Amber Glass Bottle
937796-11-7
MFCD09863240
(2-Pyrrolidin-1-ylpyrimidin-5-yl)methanol

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning