missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyrrolidin-1-ylisonicotinic acid, ≥90%, Maybridge

Chemical Identifiers

CAS 98088-04-1
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula 1·5 H2O
Formula Weight 219.25
MDL Number MFCD04115212
Chemical Name or Material 2-Pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC50401DA SDS Fisher Scientific Company
CC50401DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC50401CB SDS Fisher Scientific Company
CC50401CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

98088-04-1
100.0
1·5 H2O
MFCD04115212
90%
89.5
90%
219.25
2-Pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate

Specifications

1g
89.5
Amber Glass Bottle
1·5 H2O
MFCD04115212
90%
98088-04-1
100.0
90%
219.25
2-Pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning