missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde, 97%, Maybridge

Chemical Identifiers

CAS 900015-48-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H10N2OS
Formula Weight 182.25
MDL Number MFCD07021330
Chemical Name or Material 2-Pyrrolidin-1-yl-1, 3-thiazole-5-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC64204DA SDS Fisher Scientific Company
CC64204DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC64204CB SDS Fisher Scientific Company
CC64204CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

900015-48-7
100.0
C8H10N2OS
MFCD07021330
97%
96.5
97%
182.25
2-Pyrrolidin-1-yl-1, 3-thiazole-5-carbaldehyde

Specifications

1g
96.5
Amber Glass Bottle
C8H10N2OS
MFCD07021330
97%
900015-48-7
100.0
97%
182.25
2-Pyrrolidin-1-yl-1, 3-thiazole-5-carbaldehyde

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning