missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyridylacetic acid hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 131950250

 View more versions of this product

Catalog No. AC131950250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Pyridylacetic acid hydrochloride
16179-97-8
100.0
White to Beige
98.5% min. (Argentometry)
C7H7NO2·HCl
MFCD00012812
2-pyridylacetic acid hydrochloride, 2-pyridin-2-yl acetic acid hydrochloride, 2-pyridineacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride, pyridin-2-ylacetic acid hydrochloride, pyridylacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride 1:1, 2-pyridylacetic acid hydrochloide, pyridin-2-yl acetic acid hydrochloride, 2-2-pyridyl acetic acid hydrochloride
C1=CC=NC(=C1)CC(=O)O.Cl
173.6
173.6
Crystalline Powder or Crystals
99%
98.5
135°C
Authentic
Glass bottle
(C5H4N)CH2COOH·HCl
25 g
MQVISALTZUNQSK-UHFFFAOYSA-N
2-pyridin-2-ylacetic acid;hydrochloride
85317
99%

Chemical Identifiers

16179-97-8
173.6
MQVISALTZUNQSK-UHFFFAOYSA-N
85317
C1=CC=NC(=C1)CC(=O)O.Cl
C7H7NO2·HCl
MFCD00012812
2-pyridylacetic acid hydrochloride, 2-pyridin-2-yl acetic acid hydrochloride, 2-pyridineacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride, pyridin-2-ylacetic acid hydrochloride, pyridylacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride 1:1, 2-pyridylacetic acid hydrochloide, pyridin-2-yl acetic acid hydrochloride, 2-2-pyridyl acetic acid hydrochloride
2-pyridin-2-ylacetic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 240-316-8

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only