Please login to your online account to display your discounted pricing

2-Pyridylacetic Acid Hydrochloride 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  131950050

 View more versions of this product

Catalog No. AC131950050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder or Crystals
Formula Weight 173.6
Quantity 5g
Color White to beige
TSCA TSCA
EINECS Number 240-316-8
CAS Max % 100.0
Synonym 2-Pyridineacetic acid
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Assay Percent Range 99%
Chemical Name or Material 2-Pyridylacetic acid hydrochloride
Linear Formula (C5H4N)CH2COOH·HCl
Molecular Formula C7H7NO2·HCl
Name Note 99%
MDL Number MFCD00012812
CAS 16179-97-8

Specifications

Chemical Name or Material 2-Pyridylacetic acid hydrochloride
Name Note 99%
CAS 16179-97-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H7NO2·HCl
Linear Formula (C5H4N)CH2COOH·HCl
Formula Weight 173.6
MDL Number MFCD00012812
Synonym 2-Pyridineacetic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents