missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Pyridyl trifluoromethanesulfonate, 98%

Manufacturer:  Alfa Aesar™ H55855AA

 View more versions of this product

Catalog No. AAH55855AA



Specifications

Specifications

2-Pyridyl trifluoromethanesulfonate
1.4350
Colorless to Orange
65007-00-3
MFCD00192531
2-pyridyl triflate
C1=CC=NC(=C1)OS(=O)(=O)C(F)(F)F
227.157
227.16
98%
1.477g/mL
108°C (25mmHg)
5mL
C6H4F3NO3S
UN1993
COLRMVLTWJTLFJ-UHFFFAOYSA-N
pyridin-2-yl trifluoromethanesulfonate
3947626
Liquid

Chemical Identifiers

65007-00-3
227.157
COLRMVLTWJTLFJ-UHFFFAOYSA-N
3947626
C1=CC=NC(=C1)OS(=O)(=O)C(F)(F)F
C6H4F3NO3S
MFCD00192531
2-pyridyl triflate
pyridin-2-yl trifluoromethanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents