Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Pyridyl trifluoromethanesulfonate, 98%

Chemical Identifiers

Chemical Name or Material 2-Pyridyl trifluoromethanesulfonate
CAS 65007-00-3
Formula Weight 227.16
MDL Number MFCD00192531
UN Number UN1993
Synonym 2-Pyridyl triflate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH55855AA

  SDS
alfa aesar™
H55855AA
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55855AC

  SDS
alfa aesar™
H55855AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.16
Chemical Name or Material 2-Pyridyl trifluoromethanesulfonate
Refractive Index 1.4350
Physical Form Liquid
Assay 98%
MDL Number MFCD00192531
CAS 65007-00-3
UN Number UN1993
Synonym 2-Pyridyl triflate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density 1.477g/mL
Boiling Point 108°C (25mmHg)
Color Colorless to orange
Quantity 5mL
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.