missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyridinethioamide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC432470050

 View more versions of this product

Catalog No. AC432470050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
138.19
Yellow
96.0
Glass bottle
MFCD00087576
5346-38-3
Authentic
Crystalline Powder
5g
135° to 139°C
100.0
2-Pyridinethioamide
C6H6N2S
97%
Pyridine-2-carbothioamide

Chemical Identifiers

5346-38-3
100.0
C6H6N2S
MFCD00087576
2-Pyridinethioamide
96.0
97%
138.19
Pyridine-2-carbothioamide
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.