missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Pyridinecarboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC04001DA
Specifications
2-Pyridinecarboxylic acid | |
96.5 | |
97% | |
C6H5NO2 | |
1g | |
SIOXPEMLGUPBBT-UHFFFAOYSA-N | |
pyridine-2-carboxylic acid | |
1018 | |
123.11 |
98-98-6 | |
100.0 | |
Amber Glass Bottle | |
MFCD00006293 | |
picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique | |
C1=CC=NC(=C1)C(=O)O | |
123.111 | |
CHEBI:28747 | |
97% |
Chemical Identifiers
98-98-6 | |
123.111 | |
SIOXPEMLGUPBBT-UHFFFAOYSA-N | |
1018 | |
pyridine-2-carboxylic acid |
C6H5NO2 | |
MFCD00006293 | |
picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique | |
CHEBI:28747 | |
C1=CC=NC(=C1)C(=O)O |
Safety and Handling
GHS P Statement Harmful if swallowed. Causes serious eye irritation.
GHS P Statement Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contac
Warning