Please login to your online account to display your discounted pricing

2-Pyridinecarboxylic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC04001EA

 View more versions of this product

Catalog No. CC04001EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 123.11
Quantity 10g
Assay 97%
CAS Max % 100.0
Synonym 2-Pyridinecarboxylic acid
MDL Number MFCD00006293
Health Hazard 1 Warning
CAS 98-98-6
Molecular Formula C6H5NO2
Assay Percent Range 97%
CAS Min % 96.5
Packaging Amber Glass Bottle
Chemical Name or Material 2-Pyridinecarboxylic acid
Health Hazard 2 GHS P Statement Harmful if swallowed. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contac

Specifications

Chemical Name or Material 2-Pyridinecarboxylic acid
CAS 98-98-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H5NO2
Formula Weight 123.11
MDL Number MFCD00006293
Synonym 2-Pyridinecarboxylic acid
Accessories

Warning

GHS P Statement Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contac

GHS P Statement Harmful if swallowed. Causes serious eye irritation.

SDS & Safety Documents