Learn More
2-Pyridinecarboxaldehyde, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131820250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Pyridinecarboxaldehyde | |
98.5 | |
Brown-Yellow to Yellow | |
180.0°C to 182.0°C (761.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5343 to 1.5373 | |
MFCD00006290 | |
21, I, 287 | |
2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde | |
CSDSSGBPEUDDEE-UHFFFAOYSA-N | |
pyridine-2-carbaldehyde | |
14273 | |
107.11 | |
Liquid |
1121-60-4 | |
100.0 | |
1.1210g/mL | |
73°C | |
99% | |
C6H5NO | |
(C5H4N)COH | |
25 g | |
1.121 | |
Solubility in water: miscible. Other solubilities: soluble in most common organic solvents | |
C1=CC=NC(=C1)C=O | |
107.11 | |
CHEBI:73012 | |
99% |
Chemical Identifiers
1121-60-4 | |
107.11 | |
CSDSSGBPEUDDEE-UHFFFAOYSA-N | |
14273 | |
pyridine-2-carbaldehyde |
C6H5NO | |
MFCD00006290 | |
2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde | |
CHEBI:73012 | |
C1=CC=NC(=C1)C=O |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occur
GHS Signal Word: Danger
EINECSNumber : 214-333-6
RUO – Research Use Only