missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439890050

 View more versions of this product

Catalog No. AC439890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Pyridineboronic acid N-phenyl-diethanolamine ester
Consistent with structure
White
5g
C15H17BN2O2
Contains varying amounts of IPA and N-phenyldiethanolamine
B1(OCCN(CCO1)C2=CC=CC=C2)C3=CC=CC=N3.CC(C)O
268.13
268.13
50-70%,see CofA for exact com
Authentic
Glass bottle
>300°C
662138-96-7,67-63-0,120-07-0
2-pyridineboronic acid 2,2'-phenylimino diethanol ester, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol, pubchem6434, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol
XMHAHEGWYGUDFM-UHFFFAOYSA-N
6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane;propan-2-ol
49761666
Powder

Chemical Identifiers

662138-96-7
268.13
2-pyridineboronic acid 2,2'-phenylimino diethanol ester, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol, pubchem6434, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol
6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane;propan-2-ol
C15H17BN2O2
XMHAHEGWYGUDFM-UHFFFAOYSA-N
49761666
B1(OCCN(CCO1)C2=CC=CC=C2)C3=CC=CC=N3.CC(C)O
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves

GHS Signal Word: Warning

SDS Documents