missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC439890010
Specifications
2-Pyridineboronic acid N-phenyl-diethanolamine ester | |
Authentic | |
Glass bottle | |
>300°C | |
662138-96-7,67-63-0,120-07-0 | |
2-pyridineboronic acid 2,2'-phenylimino diethanol ester, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol, pubchem6434, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol | |
B1(OCCN(CCO1)C2=CC=CC=C2)C3=CC=CC=N3.CC(C)O | |
268.13 | |
268.13 | |
50-70%,see CofA for exact com |
Contains varying amounts of IPA and N-phenyldiethanolamine | |
Consistent with structure | |
White | |
1g | |
C15H17BN2O2 | |
XMHAHEGWYGUDFM-UHFFFAOYSA-N | |
6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane;propan-2-ol | |
49761666 | |
Powder |
Chemical Identifiers
662138-96-7 | |
268.13 | |
2-pyridineboronic acid 2,2'-phenylimino diethanol ester, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol, pubchem6434, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol | |
6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane;propan-2-ol |
C15H17BN2O2 | |
XMHAHEGWYGUDFM-UHFFFAOYSA-N | |
49761666 | |
B1(OCCN(CCO1)C2=CC=CC=C2)C3=CC=CC=N3.CC(C)O |
Safety and Handling
GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves
GHS Signal Word: Warning