missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 439890100

 View more versions of this product

Catalog No. AC439890100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Contains varying amounts of IPA and N-phenyldiethanolamine
662138-96-7,67-63-0,120-07-0
>300°C
Authentic
MFCD06798116
10g
XMHAHEGWYGUDFM-UHFFFAOYSA-N
6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane;propan-2-ol
49761666
Powder
2-Pyridineboronic acid N-phenyl-diethanolamine ester
White
C18H25BN2O3
Consistent with structure
Glass bottle
2-pyridineboronic acid 2,2'-phenylimino diethanol ester, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol, pubchem6434, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol
CC(C)O.C1CN(CCOB(O1)C1=CC=CC=N1)C1=CC=CC=C1
328.22
268.13
50-70%,see CofA for exact com

Chemical Identifiers

662138-96-7
328.22
XMHAHEGWYGUDFM-UHFFFAOYSA-N
49761666
CC(C)O.C1CN(CCOB(O1)C1=CC=CC=N1)C1=CC=CC=C1
C18H25BN2O3
MFCD06798116
2-pyridineboronic acid 2,2'-phenylimino diethanol ester, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol, pubchem6434, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol
6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane;propan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves

GHS Signal Word: Warning

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only