missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Pyridineacetic acid hydrochloride, 99%

$64.46 - $219.91

Chemical Identifiers

CAS 16179-97-8
Molecular Formula C7H7NO2·HCl
Formula Weight 173.6
MDL Number MFCD00012812
Beilstein 3696607
Synonym 2-Pyridylacetic acid hydrochloride
Chemical Name or Material 2-Pyridineacetic acid hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0487006 SDS Alfa Aesar™
L0487006
5g Each for $64.46
AAL0487014 SDS Alfa Aesar™
L0487014
25g Each for $219.91
Specifications

Description

Applications
2-Pyridineacetic acid hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

16179-97-8
173.6
3696607
2-Pyridineacetic acid hydrochloride
C7H7NO2·HCl
MFCD00012812
2-Pyridylacetic acid hydrochloride

Specifications

≈130°C
16179-97-8
173.6
3696607
2-Pyridineacetic acid hydrochloride
5g
C7H7NO2·HCl
MFCD00012812
2-Pyridylacetic acid hydrochloride
99%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA