Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Pyridineacetamide, 98%

$86.50 - $1,151.16

Chemical Identifiers

Chemical Name or Material 2-Pyridineacetamide
CAS 5451-39-8
Molecular Formula C7H8N2O
Formula Weight 136.15
MDL Number MFCD00087532
Synonym 2-(2-Pyridyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2493906

  SDS
alfa aesar™
B2493906
5g Each for $292.31

AAB2493903

  SDS
alfa aesar™
B2493903
1g Each for $86.50

AAB2493914

  SDS
alfa aesar™
B2493914
25g Each for $1,151.16
Description & Specifications

Specifications

Formula Weight 136.15
Chemical Name or Material 2-Pyridineacetamide
Assay 98%
Molecular Formula C7H8N2O
MDL Number MFCD00087532
CAS 5451-39-8
Synonym 2-(2-Pyridyl)acetamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 118° to 120°C
Quantity 5g

Applications
2-Pyridineacetamide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.