missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Pyridin-3-ylaniline, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC41814DA
Specifications
2-Pyridin-3-ylaniline | |
95% | |
C11H10N2 | |
1g | |
MTYHJHATMPCNLW-UHFFFAOYSA-N | |
2-pyridin-3-ylaniline | |
459519 | |
95% |
177202-83-4 | |
Amber Glass Bottle | |
MFCD08060516 | |
2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine | |
NC1=CC=CC=C1C1=CN=CC=C1 | |
170.22 | |
170.21 |
Chemical Identifiers
177202-83-4 | |
170.22 | |
MTYHJHATMPCNLW-UHFFFAOYSA-N | |
459519 | |
NC1=CC=CC=C1C1=CN=CC=C1 |
C11H10N2 | |
MFCD08060516 | |
2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine | |
2-pyridin-3-ylaniline |