missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2-Pyridin-3-ylaniline, 95%, Thermo Scientific™
Chemical Identifiers
CAS |
177202-83-4 |
Molecular Formula |
C11H10N2 |
Molecular Weight (g/mol) |
170.22 |
MDL Number |
MFCD08060516 |
InChI Key |
MTYHJHATMPCNLW-UHFFFAOYSA-N |
Synonym |
2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine |
PubChem CID |
459519 |
IUPAC Name |
2-pyridin-3-ylaniline |
SMILES |
NC1=CC=CC=C1C1=CN=CC=C1 |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
177202-83-4 |
|
170.22 |
|
MTYHJHATMPCNLW-UHFFFAOYSA-N |
|
459519 |
|
NC1=CC=CC=C1C1=CN=CC=C1 |
|
C11H10N2 |
|
MFCD08060516 |
|
2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine |
|
2-pyridin-3-ylaniline |
Specifications
|
177202-83-4 |
|
Amber Glass Bottle |
|
MFCD08060516 |
|
2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine |
|
NC1=CC=CC=C1C1=CN=CC=C1 |
|
170.22 |
|
170.21 |
|
2-Pyridin-3-ylaniline |
|
95% |
|
C11H10N2 |
|
250mg |
|
MTYHJHATMPCNLW-UHFFFAOYSA-N |
|
2-pyridin-3-ylaniline |
|
459519 |
|
95% |
SDS
missing translation for 'documents'