Please login to your online account to display your discounted pricing

2-Pyridin-3-ylaniline, 95%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC41814DE

 View more versions of this product

Catalog No. CC41814DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 170.21
Quantity 5g
Assay 95%
Packaging Amber Glass Bottle
Chemical Name or Material 2-Pyridin-3-ylaniline
CAS 177202-83-4
Molecular Formula C11H10N2
Assay Percent Range 95%

Specifications

Chemical Name or Material 2-Pyridin-3-ylaniline
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H10N2
Formula Weight 170.21