Please login to your online account to display your discounted pricing

2-pyrazinecarbaldehyde, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC03404CB

 View more versions of this product

Catalog No. CC03404CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Amber Glass Bottle
Quantity 250mg
Chemical Name or Material 2-pyrazinecarbaldehyde
CAS 5780-66-5
Assay 95%
Molecular Formula C5H4N2O
Formula Weight 108.10g/mol

Specifications

Chemical Name or Material 2-pyrazinecarbaldehyde
Assay 95%
Molecular Formula C5H4N2O
Formula Weight 108.10g/mol