Please login to your online account to display your discounted pricing

2-pyrazinecarbaldehyde, Maybridge

Chemical Identifiers

Chemical Name or Material 2-pyrazinecarbaldehyde
Assay 95%
Molecular Formula C5H4N2O
Formula Weight 108.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC03404CB

  SDS
fisher scientific company
CC03404CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC03404DA

  SDS
fisher scientific company
CC03404DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.1
Quantity 250mg
Assay 95%
CAS 5780-66-5
Chemical Name or Material 2-pyrazinecarbaldehyde
Packaging Amber Glass Bottle
Molecular Formula C5H4N2O