missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propylthiazole, TCI America™

Manufacturer:  TCI America P183725G

 View more versions of this product

Catalog No. P183725G



Specifications

Specifications

2-Propylthiazole
Yellow
17626-75-4
MFCD00053154
2-propylthiazole, thiazole, 2-propyl, 2-n-propylthiazole, unii-t395979vrl, 2-propylthiazol, thiazole,2-propyl, 2-normal-propyl thiazole, acmc-209eb6, #
CCCC1=NC=CS1
127.205
127.21
≥98.0% (GC)
172°C
25g
C6H9NS
1993
CMOIEFFAOUQJPS-UHFFFAOYSA-N
2-propyl-1,3-thiazole
87198
Clear Liquid at 20°C

Chemical Identifiers

17626-75-4
127.205
CMOIEFFAOUQJPS-UHFFFAOYSA-N
87198
CCCC1=NC=CS1
C6H9NS
MFCD00053154
2-propylthiazole, thiazole, 2-propyl, 2-n-propylthiazole, unii-t395979vrl, 2-propylthiazol, thiazole,2-propyl, 2-normal-propyl thiazole, acmc-209eb6, #
2-propyl-1,3-thiazole
Safety and Handling

Safety and Handling

TSCA : Yes

Documents