missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propoxybenzoic Acid, TCI America™

Manufacturer:  TCI America P169425G

 View more versions of this product

Catalog No. P169425G



Specifications

Specifications

2-Propoxybenzoic Acid
Undesignated
25g
C10H12O3
2-n-propoxybenzoic acid, o-propoxybenzoic acid, benzoic acid, 2-propoxy, 2-propoxy benzoic acid, salicylic acid propyl ether, akos bbb/270, rarechem al bo 0876, propyloxybenzoic acid, 2-propoxybenzoic aci, o-propoxy benzoic acid
CCCOC1=CC=CC=C1C(=O)O
180.203
180.20
≥98.0% (GC,T)
207°C
39°C
2100-31-4
MFCD01075687
OXOWWPXTTOCKKU-UHFFFAOYSA-N
2-propoxybenzoic acid
539215
Crystal-Powder at 20°C

Chemical Identifiers

2100-31-4
180.203
OXOWWPXTTOCKKU-UHFFFAOYSA-N
539215
CCCOC1=CC=CC=C1C(=O)O
C10H12O3
MFCD01075687
2-n-propoxybenzoic acid, o-propoxybenzoic acid, benzoic acid, 2-propoxy, 2-propoxy benzoic acid, salicylic acid propyl ether, akos bbb/270, rarechem al bo 0876, propyloxybenzoic acid, 2-propoxybenzoic aci, o-propoxy benzoic acid
2-propoxybenzoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents