missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Propionylthiophene, 98%, Thermo Scientific Chemicals
$104.58 - $266.66
Chemical Identifiers
CAS | 13679-75-9 |
---|---|
Molecular Formula | C7H8OS |
Molecular Weight (g/mol) | 140.2 |
MDL Number | MFCD00005446 |
InChI Key | MFPZQZZWAMAHOY-UHFFFAOYSA-N |
Synonym | 1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone |
PubChem CID | 26179 |
IUPAC Name | 1-thiophen-2-ylpropan-1-one |
SMILES | CCC(=O)C1=CC=CS1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0231814
|
Thermo Scientific Chemicals
L0231814 |
25 g |
Each for $104.58
|
|
|||||
AAL0231822
|
Thermo Scientific Chemicals
L0231822 |
100 g |
Each for $266.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13679-75-9 | |
140.2 | |
MFPZQZZWAMAHOY-UHFFFAOYSA-N | |
26179 | |
CCC(=O)C1=CC=CS1 |
C7H8OS | |
MFCD00005446 | |
1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone | |
1-thiophen-2-ylpropan-1-one |
Specifications
13679-75-9 | |
228°C | |
C7H8OS | |
MFCD00005446 | |
108934 | |
MFPZQZZWAMAHOY-UHFFFAOYSA-N | |
1-thiophen-2-ylpropan-1-one | |
26179 | |
98% |
1.124 | |
100°C (212°F) | |
1.553 | |
25 g | |
1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone | |
CCC(=O)C1=CC=CS1 | |
140.2 | |
140.2 | |
2-Propionylthiophene |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 237-182-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only