missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propionylthiazole, TCI America™

Manufacturer:  TCI America P22475G

 View more versions of this product

Catalog No. P22475G



Specifications

Specifications

2-Propionylthiazole
Yellow
43039-98-1
MFCD01681404
TYRAENAWSLPSLW-UHFFFAOYSA-N
1-(1,3-thiazol-2-yl)propan-1-one
65288
Clear Liquid at 20°C
107°C
5g
C6H7NOS
2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k
CCC(=O)C1=NC=CS1
141.188
141.19
≥98.0% (GC)

Chemical Identifiers

43039-98-1
141.188
TYRAENAWSLPSLW-UHFFFAOYSA-N
65288
CCC(=O)C1=NC=CS1
C6H7NOS
MFCD01681404
2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k
1-(1,3-thiazol-2-yl)propan-1-one
Safety and Handling

Safety and Handling

RTECSNumber : XJ5123000

TSCA : Yes

Documents