missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Propionylthiazole, 99%, Thermo Scientific Chemicals
$103.19 - $345.96
Chemical Identifiers
CAS | 43039-98-1 |
---|---|
Molecular Formula | C6H7NOS |
Molecular Weight (g/mol) | 141.188 |
MDL Number | MFCD01681404 |
InChI Key | TYRAENAWSLPSLW-UHFFFAOYSA-N |
Synonym | 2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k |
PubChem CID | 65288 |
IUPAC Name | 1-(1,3-thiazol-2-yl)propan-1-one |
SMILES | CCC(=O)C1=NC=CS1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2171104
|
Thermo Scientific Chemicals
B2171104 |
2 g |
Each for $103.19
|
|
|||||
AAB2171109
|
Thermo Scientific Chemicals
B2171109 |
10 g |
Each for $345.96
|
|
|||||
Description
Used as flavoring agent and fragrance chemical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as flavoring agent and fragrance chemical compounds.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
43039-98-1 | |
141.188 | |
TYRAENAWSLPSLW-UHFFFAOYSA-N | |
65288 | |
CCC(=O)C1=NC=CS1 |
C6H7NOS | |
MFCD01681404 | |
2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k | |
1-(1,3-thiazol-2-yl)propan-1-one |
Specifications
43039-98-1 | |
110°C (5 mmHg) | |
C6H7NOS | |
2 g | |
Not miscible or difficult to mix with water. | |
CCC(=O)C1=NC=CS1 | |
141.188 | |
141.19 | |
2-Propionylthiazole |
1.174 | |
>100°C (212°F) | |
MFCD01681404 | |
2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k | |
TYRAENAWSLPSLW-UHFFFAOYSA-N | |
1-(1,3-thiazol-2-yl)propan-1-one | |
65288 | |
99% |
Safety and Handling
RTECSNumber : XJ5123000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only