missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Propargyloxy)benzaldehyde, TCI America™

Manufacturer:  TCI America P2338200MG

 View more versions of this product

Catalog No. P2338200MG



Specifications

Specifications

2-(Propargyloxy)benzaldehyde
70°C
29978-83-4
MFCD02380378
PEERMPFPCWSJLC-UHFFFAOYSA-N
2-prop-2-ynoxybenzaldehyde
583037
Crystal-Powder at 20°C
Undesignated
200mg
C10H8O2
2-(2-Propynyloxy)benzaldehyde
C#CCOC1=CC=CC=C1C=O
160.172
160.17
≥98.0% (GC)

Chemical Identifiers

29978-83-4
160.172
PEERMPFPCWSJLC-UHFFFAOYSA-N
583037
C#CCOC1=CC=CC=C1C=O
C10H8O2
MFCD02380378
2-(2-Propynyloxy)benzaldehyde
2-prop-2-ynoxybenzaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents