missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propanol, Semiconductor Grade, 99.5% min, Alfa Aesar

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Solubility in H2O P.T., H2O (by Karl Fischer titrn) 0.05%, Acidity 0.2meq/g, Alkalinity 0.1meq/g, Cl 0.2ppm, PO4 0.5ppm

Manufacturer:  Alfa Aesar 19397M6

 View more versions of this product

Catalog No. AA19397M6



Specifications

Specifications

Colorless liquid
0.785g/cm3 at 20°C
Alcohol-like
Colorless
67-63-0
99.5% min.
MFCD00011674
2-Propanol
CC(C)O
60.096
CHEBI:17824
Liquid
2-Propanol
455°C (851°F)
48 hPa (36mm Hg at 20°C)
4 x 4L
Semiconductor
C3H8O
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
Safety and Handling

Safety and Handling

H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 200-661-7

SDS Documents