Please login to your online account to display your discounted pricing

2-Propanol, PHOTREX™ Reagent, J.T.Baker™

High purity solvents characterized for use in UV, visible and IR spectrophotometry

$126.84 - $616.32

Chemical Identifiers

Chemical Name or Material 2-Propanol
CAS 67-63-0
Purity Grade PHOTREX™ Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula CH3CHOHCH3
Formula Weight 60.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002090

 
avantor performance
908303
Narrow Mouth Amber Glass Bottle 4L Each for $616.32

02002098

 
avantor performance
908301
Narrow Mouth Amber Glass Bottle 500mL Each for $126.84
Description & Specifications

Specifications

Formula Weight 60.1
Purity Grade Notes Meets ACS reagent chemical requirements
Chemical Name or Material 2-Propanol
Molecular Formula CH3CHOHCH3
CAS 67-63-0
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Purity Grade PHOTREX™ Reagent
Flash Point 12.00°C
Quantity 4L

    For U.V. Spectrophotometry; For Laboratory, Research, or Manufacturing Use