Please login to your online account to display your discounted pricing

Isopropanol (IPA), Optima™, Fisher Chemical

High purity mobile phase for HPLC and extraction solvent

$160.77 - $421.23

Chemical Identifiers

Chemical Name or Material 2-Propano
CAS 67-63-0
Purity Grade OPTIMA™
Purity Grade Notes High purity mobile phase for HPLC and/or extraction solvent
Assay ≥99.9%
Assay Percent Range 99.9% min by GC
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
Formula Weight 60.01
MDL Number MFCD00011674
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A464SK-4

  SDS
fisher scientific company
A464SK4
Amber Glass Bottle 4L Each for $421.23
Case of 4 Each for $1,684.92

A464-4

  Accessories SDS
fisher scientific company
BPA464-4
Amber Glass Bottle 4L Each for $261.31
Case of 4 Each for $1,045.24

A4641

  SDS
fisher scientific company
A4641
Amber Glass Bottle 1L Each for $160.77
Case of 6 Each for $964.62

A464RS200

  SDS
fisher scientific company
A464RS200
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Propanol
Cleanup Symbol Solvent
Absorbance 0.015 AU max. at 254nm
CAS 67-63-0
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Acetone 0.002% max.
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0
DOT Information DOT Class 3, : Flammable Liquid
Additional Information Vapor Pressure: 43mmHg at 20°C
Purity Grade OPTIMA™
Purity Grade Notes High purity mobile phase for HPLC and/or extraction solvent
Physical Form Liquid
Assay ≥99.9%
Assay Percent Range Assay: 99.9% min by GC
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
Synonym IPA
Density 0.785g/cm³
Fluorescence Background 1 ppb max.
Identification Pass Test
Packaging Amber Glass Bottle
Propionaldehyde 0.002% max.
Refractive Index 1.3740 to 1.3760 (at 25°C)
Residue after Evaporation 1ppm max.
Solubility in Water Pass Test
Substances Reducing Permanganate Pass Test
Titratable Acid or Base 0.0001 max.
Vapor Pressure 43mmHg at 20°C
Water 0.05% max.
Viscosity 2.27 mPa/s at 20°C
Alpha Vector Propanol
Boiling Point 81°C
Color Colorless
Melting Point −89.5°C
pH 7
Quantity 4L

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Also meets ACS specifications
  • Intended for research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!