Please login to your online account to display your discounted pricing

2-Propanol (Laboratory), Fisher Chemical

General purpose laboratory

$279.85 - $1,014.85

Chemical Identifiers

Chemical Name or Material 2-Propanol
CAS 67-63-0
Purity Grade Laboratory Grade
Purity Grade Notes Purified
Physical Form Liquid
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
Synonym Isopropanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A415-20

  SDS
fisher scientific company
BPA41520
Steel Pail 20L Each for $1,014.85

A415-4

  Accessories SDS
fisher scientific company
BPA4154
Glass Bottle 4L Each for $279.85
Case of 4 Each for $1,119.40
Description & Specifications

Specifications

Melting Point −89.5°C
Purity Grade Laboratory Grade
Boiling Point 81°C
Formula Weight 60.1
Quantity 20L
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPa/s at 20°C
Alpha Vector Propanol
Physical Form Liquid
Color Colorless
pH 7
Density 0.785g/cm³
Identification Pass Test
Additional Information Vapor Pressure: 43mmHg at 20°C
Packaging Steel Pail
Purity Grade Notes Purified
MDL Number MFCD00011674
Synonym Isopropanol
Health Hazard 2 WARNING!
Cleanup Symbol Solvent
Linear Formula (CH3)2CHOH
ChemAlert Storage Symbol Red
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
Chemical Name or Material 2-Propanol
Molecular Formula C3H8O
CAS 67-63-0

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

WARNING!