missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propanol, HPLC Grade, 99.7+%, Alfa Aesar

Maximum level of impurities: Evaporation residue 3ppm, H2O 0.05%, Chlorinated pesticides 5ppt

Manufacturer:  Alfa Aesar 22906M6

 View more versions of this product

Catalog No. AA22906M6



Specifications

Specifications

Colorless liquid
0.785g/cm3
455°C (851°F)
82°C to 83°C
-90°C
67-63-0
C3H8O
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
Liquid
2-Propanol
12°C (53.6°F)
Alcohol-like
Colorless
4 x 4L
>99.7%
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
HPLC

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
Safety and Handling

Safety and Handling

H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 200-661-7

SDS Documents