Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Propanol-d8, 99%(Isotopic)

Chemical Identifiers

Chemical Name or Material 2-Propanol-d8
CAS 22739-76-0
Name Note Isotopic
Formula Weight 68.15
MDL Number MFCD00044341
UN Number UN1219
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4231414

  SDS
alfa aesar™
4231414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 68.15
UN Number UN1219
Melting Point −89.5°C
Physical Form Liquid
Quantity 25g
Flash Point −11.7°C (53°F)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Boiling Point 82.4°C
Odor Alcohol-like
Density 0.90g/mL
Assay 99%
Chemical Name or Material 2-Propanol-d8
Name Note Isotopic
MDL Number MFCD00044341
CAS 22739-76-0
Health Hazard 2 GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.