Please login to your online account to display your discounted pricing

2-Propanol (Certified), Fisher Chemical

$168.38 - $344.23

Chemical Identifiers

Chemical Name or Material 2-Propanol
CAS 67-63-0
Purity Grade Certified Grade
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
Synonym Isopropanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A417-1

  Accessories SDS
fisher scientific company
BPA4171
Glass Bottle 1L Each for $168.38
Case of 6 Each for $1,010.28

A417-4

  Accessories SDS
fisher scientific company
BPA4174
Glass Bottle 4L Each for $344.23
Case of 4 Each for $1,376.92
Description & Specifications

Specifications

Formula Weight 60.1
Boiling Point 81°C
Purity Grade Certified Grade
Melting Point −89.5°C
Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Density 0.785g/cm³
Vapor Pressure 43mmHg at 20°C
pH 7
Alpha Vector Propanol
Vapor Density 2.1g/mol at 20°C/ 68°F
Color Colorless
Viscosity 2.27 mPa/s at 20°C
Synonym Isopropanol
MDL Number MFCD00011674
Cleanup Symbol Solvent
Additional Information Vapor Pressure: 43mmHg at 20°C
Packaging Glass Bottle
Triglycerides Use Pass Test
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
ChemAlert Storage Symbol Red
Molecular Formula C3H8O
Chemical Name or Material 2-Propanol
CAS 67-63-0
Linear Formula (CH3)2CHOH

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

WARNING!