missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propanol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  Avantor Performance 908422

 View more versions of this product

Catalog No. 02002086



Description

Description

    For Laboratory,Research,or Manufacturing Use
Specifications

Specifications

2-Propanol
Meets ACS reagent chemical requirements
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
BAKER ANALYZED™ ACS Reagent
1L = 0.79kg
1L
67-63-0
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
Aluminum Bottle
12.00°C

Chemical Identifiers

67-63-0
60.096
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
Documents