Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Propanol, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material 2-Propanol
CAS 67-63-0
Name Note Anhydrous
Linear Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
UN Number UN1219
Beilstein 635639
Merck Index 14,5208
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41840K2

  SDS
alfa aesar™
41840K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41840AK

  SDS
alfa aesar™
41840AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.1
Odor Alcohol-like
Chemical Name or Material 2-Propanol
Beilstein 635639
Boiling Point 82° to 83°C
RTECS Number NT8050000
Physical Form Liquid
MDL Number MFCD00011674
Refractive Index 1.3770
Packaging ChemSeal™ bottles
Merck Index 14,5208
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
UN Number UN1219
Health Hazard 2 GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Name Note Anhydrous
Quantity 1L
CAS 67-63-0
Storage Note 1
  • Packaged under argon
Flash Point 12° (53°F)
Health Hazard 1 Danger
Color Colorless
Melting Point −90°C
TSCA TSCA
Assay ≥99.5%
Linear Formula (CH3)2CHOH
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 0.785g/mL

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

  • Packaged under argon