Please login to your online account to display your discounted pricing

2-Propanol, ACS, 99.5% min, Alfa Aesar™

Solvent for gums, resins, alkaloids, essential oils. Denaturant for ethyl alcohol. In antifreeze compositions

Chemical Identifiers

Chemical Name or Material 2-Propanol
CAS 67-63-0
Purity Grade ACS Grade
Molecular Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
UN Number UN1219
Beilstein 635639
Merck Index 14,5208
Synonym IPA; Isopropanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA36644M1

 
alfa aesar
36644M1
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 60.1
Quantity 4x1L
Chemical Name or Material 2-Propanol
TSCA TSCA
Odor Alcohol-like
Refractive Index 1.3770
Flash Point 12°C (53°F)
Boiling Point 82° to 83°C
Physical Form Liquid
Melting Point −90°C
Assay ≥99.5%
Molecular Formula (CH3)2CHOH
MDL Number MFCD00011674
CAS 67-63-0
UN Number UN1219
Beilstein 635639
Merck Index 14,5208
Synonym IPA; Isopropanol
Solubility Information Miscible with water, alcohol, ether, chloroform
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H319-H336;
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Density 0.785
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
RTECS Number NT8050000
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319-H336;
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.