Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Propanol, 99+%

Chemical Identifiers

Chemical Name or Material 2-Propanol
CAS 67-63-0
Molecular Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
UN Number UN1219
Beilstein 635639
Merck Index 14,5208
Synonym Isopropanol; Isopropyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL101810F

  SDS
alfa aesar™
L101810F
2500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.1
Chemical Name or Material 2-Propanol
Assay ≥99%
Health Hazard 2 GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 1 Danger
Beilstein 635639
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
MDL Number MFCD00011674
CAS 67-63-0
UN Number UN1219
Boiling Point 82° to 83°C
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 2500mL
Synonym Isopropanol; Isopropyl alcohol
Melting Point −90°C
RTECS Number NT8050000
Odor Alcohol-like
Merck Index 14,5208
Flash Point 12°C (53°F)
TSCA TSCA
Molecular Formula (CH3)2CHOH

Applications
2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.

Solubility
Miscible with water, ethanol, acetone, chloroform, methanol, diethyl ether, n-octanol and benzene.

Notes
It is a flammable chemical compound and is hygroscopic in nature. Heat and sunlight can contribute to its instability.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.