Learn More
2-Propanethiol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131445000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Propanethiol | |
75-33-2 | |
100.0 | |
0.8200g/mL | |
-34°C | |
97.5% min. (GC) | |
C3H8S | |
(CH3)2CHSH | |
500 mL | |
0.82 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohols,ethyl ether,hydrocarbons | |
CC(C)S | |
76.15 | |
CHEBI:8474 | |
98% |
98% | |
97.5 | |
-131°C | |
57°C to 60°C | |
Authentic | |
Glass bottle | |
1.4245 to 1.4265 | |
MFCD00004863 | |
01, 367 | |
2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol | |
KJRCEJOSASVSRA-UHFFFAOYSA-N | |
propane-2-thiol | |
6364 | |
76.15 | |
Liquid |
Chemical Identifiers
75-33-2 | |
76.15 | |
KJRCEJOSASVSRA-UHFFFAOYSA-N | |
6364 | |
propane-2-thiol |
C3H8S | |
MFCD00004863 | |
2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol | |
CHEBI:8474 | |
CC(C)S |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation or rash occurs: Get medical advice/attention.
Keep away from heat/sparks/o
GHS Signal Word: Danger
EINECSNumber : 200-861-4
RTECSNumber : TZ7302000
TSCA : TSCA
RUO – Research Use Only