missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propanethiol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 043422AP

 View more versions of this product

Catalog No. AA43422AP



Description

Description

2-Propanethiol Reacts with 3-chloro-propene to get 3-isopropylsulfanyl-propene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Specifications

2-Propanethiol
-131°C
0.82 g/mL
-34° (-29°F)
C3H8S
(CH3)2CHSH
500 mL
605260
2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol
KJRCEJOSASVSRA-UHFFFAOYSA-N
propane-2-thiol
6364
76.16
Liquid
75-33-2
Colorless
57°C to 60°C
Stench
1.4255
MFCD00004863
UN2402
Air Sensitive
Slightly soluble in water.
CC(C)S
76.157
CHEBI:8474
98%

Chemical Identifiers

75-33-2
76.157
KJRCEJOSASVSRA-UHFFFAOYSA-N
6364
propane-2-thiol
C3H8S
MFCD00004863
2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol
CHEBI:8474
CC(C)S
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P261-P272-P280-P303+P361+P353-P333+P313-P363-P370+P378q-P501c

H225-H317

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPANETHIOLS

EINECSNumber : 200-861-4

RTECSNumber : TZ7302000

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only