Please login to your online account to display your discounted pricing

2-Propanesulfonic Acid Sodium Salt, Monohydrate 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  418350100

 View more versions of this product

Catalog No. AC418350100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 164.16
Quantity 10g
Color White
EINECS Number 226-434-2
TSCA TSCA
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Propanesulfonic acid, sodium salt monohydrate
Name Note 99%
Infrared Spectrum Authentic
Molecular Formula C3 H7 Na O3 S . H2 O
Packaging Glass bottle
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Additional Information sodium bromide content (by titr.): < 1%
Assay Percent Range 99%
Linear Formula (CH3)2CHSO3Na·H2O
MDL Number MFCD00150629
CAS 5399-58-6

Specifications

Chemical Name or Material 2-Propanesulfonic acid, sodium salt monohydrate
Name Note 99%
CAS 5399-58-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3 H7 Na O3 S . H2 O
Linear Formula (CH3)2CHSO3Na·H2O
Formula Weight 164.16
MDL Number MFCD00150629
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents