Please login to your online account to display your discounted pricing

2-Propanesulfonic Acid Sodium Salt, Monohydrate 99%, ACROS Organics™

$297.81 - $297.81

Chemical Identifiers

Chemical Name or Material 2-Propanesulfonic acid, sodium salt monohydrate
Name Note 99%
CAS 5399-58-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3 H7 Na O3 S . H2 O
Linear Formula (CH3)2CHSO3Na·H2O
Formula Weight 164.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC418350100

  SDS
thermo fisher scientific
418350100
Glass bottle 10g Each for $297.81
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 164.16
Physical Form Crystalline Powder
Quantity 10g
Color White
TSCA TSCA
EINECS Number 226-434-2
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 98.5
Additional Information sodium bromide content (by titr.): < 1%
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material 2-Propanesulfonic acid, sodium salt monohydrate
Name Note 99%
Molecular Formula C3 H7 Na O3 S . H2 O
CAS 5399-58-6
Assay Percent Range 99%
MDL Number MFCD00150629
Linear Formula (CH3)2CHSO3Na·H2O
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.