Please login to your online account to display your discounted pricing

2-PMDQ, Tocris

Potent, selective α1 antagonist

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material 2-PMDQ
Assay Percent Range >98%
Molecular Formula C21H23N5O
Formula Weight 361.45
Synonym 2-[(4-Phenylpiperazin-1-yl)methyl]-2, 3-dihydroimidazo[1, 2-c]quinazolin-5(6H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

062710

 
r & d systems
062710
10mg Each for $195.49

062750

 
r & d systems
062750
50mg Each for $816.90
Description & Specifications

Specifications

Chemical Name or Material 2-PMDQ
Quantity 10mg
CAS 139047-55-5
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C21H23N5O
Formula Weight 361.45
Synonym 2-[(4-Phenylpiperazin-1-yl)methyl]-2, 3-dihydroimidazo[1, 2-c]quinazolin-5(6H)-one
Safety and Handling

Store at Room Temperature