Please login to your online account to display your discounted pricing

2-PMDQ, Tocris


Potent, selective α1 antagonist

Quantity 50mg

Manufacturer: r & d systems  062750

 View more versions of this product

Catalog No. 062750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 361.45
Assay Percent Range >98%
Synonym 2-[(4-Phenylpiperazin-1-yl)methyl]-2, 3-dihydroimidazo[1, 2-c]quinazolin-5(6H)-one
Chemical Name or Material 2-PMDQ
CAS 139047-55-5
Molecular Formula C21H23N5O
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material 2-PMDQ
Assay Percent Range >98%
Molecular Formula C21H23N5O
Formula Weight 361.45
Synonym 2-[(4-Phenylpiperazin-1-yl)methyl]-2, 3-dihydroimidazo[1, 2-c]quinazolin-5(6H)-one
Accessories

Store at Room Temperature