Please login to your online account to display your discounted pricing

2-(Piperidylmethyl)-1-cyclohexanone hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Piperidylmethyl)-1-cyclohexanone hydrochloride
Name Note 98%
CAS 1850565
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C12H21NO·HCl
Formula Weight 231.77
MDL Number MFCD00068799
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC335630010

  SDS
thermo fisher scientific
335630010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.77
Quantity 1g
Merck Index 13, 7510
RTECS Number GW2080000
TSCA TSCA
CAS Max % 100.0
CAS 1850565
Packaging Glass bottle
CAS Min % 97.5
Molecular Formula C12H21NO·HCl
Name Note 98%
Chemical Name or Material 2-(Piperidylmethyl)-1-cyclohexanone hydrochloride
Assay Percent Range 98%
MDL Number MFCD00068799