missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Piperidinopyrid-4-yl)methylamine, 97%, Maybridge

Chemical Identifiers

CAS 876316-37-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H17N3
Formula Weight 191.28
MDL Number MFCD08690284
Chemical Name or Material (2-Piperidinopyrid-4-yl)methylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC50313EA SDS Fisher Scientific Company
CC50313EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

876316-37-9
100.0
C11H17N3
MFCD08690284
97%
96.5
97%
191.28
(2-Piperidinopyrid-4-yl)methylamine

Specifications

191.28
100.0
MFCD08690284
876316-37-9
Amber Glass Bottle
97%
10g
C11H17N3
97%
96.5
(2-Piperidinopyrid-4-yl)methylamine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger