missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Piperidinoisonicotinic acid, 97%, Maybridge

Chemical Identifiers

CAS 855153-75-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H14N2O2
Formula Weight 206.24
MDL Number MFCD04115214
Chemical Name or Material 2-Piperidinoisonicotinic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC50301CB SDS Fisher Scientific Company
CC50301CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC50301DA SDS Fisher Scientific Company
CC50301DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

855153-75-2
100.0
C11H14N2O2
MFCD04115214
97%
96.5
97%
206.24
2-Piperidinoisonicotinic acid

Specifications

206.24
100.0
97%
C11H14N2O2
MFCD04115214
97%
250mg
Amber Glass Bottle
855153-75-2
96.5
2-Piperidinoisonicotinic acid

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning