missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-piperidinoaniline, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company BTB02217EA

 View more versions of this product

Catalog No. BTB02217EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Piperidinoaniline
10g
96.5
97%
MFCD00047467
OYECAJPUPWFCSL-UHFFFAOYSA-N
2-piperidin-1-ylaniline
458795
97%
Amber Glass Bottle
39643-31-7
100.0
C11H16N2
1-2-aminophenyl piperidine, 2-1-piperidino aniline, 2-1-piperidinyl aniline, 2-piperdinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, 2-piperidinoaniline, 2-piperidylphenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine
NC1=CC=CC=C1N1CCCCC1
176.26
176.26

Chemical Identifiers

39643-31-7
176.26
OYECAJPUPWFCSL-UHFFFAOYSA-N
458795
NC1=CC=CC=C1N1CCCCC1
C11H16N2
MFCD00047467
1-2-aminophenyl piperidine, 2-1-piperidino aniline, 2-1-piperidinyl aniline, 2-piperdinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, 2-piperidinoaniline, 2-piperidylphenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine
2-piperidin-1-ylaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents